Anti-MPP5 Mouse Monoclonal Antibody [clone: 5D10]
Leverandør: OriGene
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ORIGCF504458EA 5750 DKK
ORIGCF504458
Anti-MPP5 Mouse Monoclonal Antibody [clone: 5D10]
Antistoffer
Recommended Dilutions: Western Blot: 1:500-2000
Type: Primary
Antigen: MPP5
Clonality: Monoclonal
Clone: 5D10
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human