Anti-PPAT Mouse Monoclonal Antibody [clone: 3C6]
Leverandør: OriGene
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ORIGCF504457EA 5590 DKK
ORIGCF504457
Anti-PPAT Mouse Monoclonal Antibody [clone: 3C6]
Antistoffer
Recommended Dilutions: Western Blot: 1:200-1:1000
Type: Primary
Antigen: PPAT
Clonality: Monoclonal
Clone: 3C6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human