Anti-SERPINB13 Mouse Monoclonal Antibody [clone: 1A7]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF503707EA 5590 DKK
ORIGCF503707
Anti-SERPINB13 Mouse Monoclonal Antibody [clone: 1A7]
Antistoffer
Recommended Dilutions: Western Blot: 1:500-2000; Flow Cytometry: 1:100

Type: Primary
Antigen: SERPINB13
Clonality: Monoclonal
Clone: 1A7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Monkey, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR