Anti-ZNF397 Mouse Monoclonal Antibody [clone: 1F3]
Leverandør: OriGene
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ORIGCF503687EA 5590 DKK
ORIGCF503687
Anti-ZNF397 Mouse Monoclonal Antibody [clone: 1F3]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100
Type: Primary
Antigen: ZNF397
Clonality: Monoclonal
Clone: 1F3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human