Anti-DAND5 Mouse Monoclonal Antibody [clone: 3D4]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF503675EA 5590 DKK
ORIGCF503675
Anti-DAND5 Mouse Monoclonal Antibody [clone: 3D4]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: DAND5
Clonality: Monoclonal
Clone: 3D4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR