Anti-MAP2K3 Mouse Monoclonal Antibody [clone: 6F8]
Leverandør: OriGene
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ORIGCF505643EA 5590 DKK
ORIGCF505643
Anti-MAP2K3 Mouse Monoclonal Antibody [clone: 6F8]
Antistoffer
Recommended Dilutions: Western Blot: 1:200-4000; Immunochemistry: 1:150; Immunofluorescence: 1:100
Type: Primary
Antigen: MAP2K3
Clonality: Monoclonal
Clone: 6F8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Dog, Human, Monkey, Mouse, Rat