Anti-CA12 Mouse Monoclonal Antibody [clone: 1A6]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF505628EA 5590 DKK
ORIGCF505628
Anti-CA12 Mouse Monoclonal Antibody [clone: 1A6]
Antistoffer
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:150

Type: Primary
Antigen: CA12
Clonality: Monoclonal
Clone: 1A6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR