Anti-ROMO1 Mouse Monoclonal Antibody [clone: 5G8]
Leverandør: OriGene
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ORIGCF505611EA 5590 DKK
ORIGCF505611
Anti-ROMO1 Mouse Monoclonal Antibody [clone: 5G8]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100
Type: Primary
Antigen: ROMO1
Clonality: Monoclonal
Clone: 5G8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human