Anti-NDEL1 Mouse Monoclonal Antibody [clone: 1G7]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF503254EA 5590 DKK
ORIGCF503254
Anti-NDEL1 Mouse Monoclonal Antibody [clone: 1G7]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: NDEL1
Clonality: Monoclonal
Clone: 1G7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Dog, Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR