Anti-C1orf50 Mouse Monoclonal Antibody [clone: 1E1]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF503252EA 5750 DKK
ORIGCF503252
Anti-C1orf50 Mouse Monoclonal Antibody [clone: 1E1]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: C1orf50
Clonality: Monoclonal
Clone: 1E1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR