Anti-CD4 Mouse Monoclonal Antibody [clone: 3B4]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF500482EA 5750 DKK
ORIGCF500482
Anti-CD4 Mouse Monoclonal Antibody [clone: 3B4]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: 3B4
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR