Anti-SHPRH Mouse Monoclonal Antibody [clone: 4A3]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF501448EA 5750 DKK
ORIGCF501448
Anti-SHPRH Mouse Monoclonal Antibody [clone: 4A3]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: SHPRH
Clonality: Monoclonal
Clone: 4A3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR