Anti-ACO2 Mouse Monoclonal Antibody [clone: 3G8]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF500824EA 5590 DKK
ORIGCF500824
Anti-ACO2 Mouse Monoclonal Antibody [clone: 3G8]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100

Type: Primary
Antigen: ACO2
Clonality: Monoclonal
Clone: 3G8
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR