Anti-DCXR Mouse Monoclonal Antibody [clone: 4D11]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF500820EA 5590 DKK
ORIGCF500820
Anti-DCXR Mouse Monoclonal Antibody [clone: 4D11]
Antistoffer
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:50; Immunofluorescence: 1:100 Flow Cytometry: 1:100

Type: Primary
Antigen: DCXR
Clonality: Monoclonal
Clone: 4D11
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR