Anti-DCXR Mouse Monoclonal Antibody [clone: 4H10]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF500819EA 5590 DKK
ORIGCF500819
Anti-DCXR Mouse Monoclonal Antibody [clone: 4H10]
Antistoffer
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:50 Flow Cytometry: 1:100

Type: Primary
Antigen: DCXR
Clonality: Monoclonal
Clone: 4H10
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR