Anti-HSD17B10 Mouse Monoclonal Antibody [clone: 5E2]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF500813EA 5590 DKK
ORIGCF500813
Anti-HSD17B10 Mouse Monoclonal Antibody [clone: 5E2]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100 Flow Cytometry: 1:100

Type: Primary
Antigen: HSD17B10
Clonality: Monoclonal
Clone: 5E2
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Monkey
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR