Anti-ID3 Mouse Monoclonal Antibody [clone: 12C7]
Leverandør: OriGene
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ORIGCF500790EA 5590 DKK
ORIGCF500790
Anti-ID3 Mouse Monoclonal Antibody [clone: 12C7]
Antistoffer
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:50; Immunofluorescence: 1:100; Flow Cytometry: 1:100
Type: Primary
Antigen: ID3
Clonality: Monoclonal
Clone: 12C7
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human