Anti-BTRC Mouse Monoclonal Antibody [clone: 1F10]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF502463EA 5590 DKK
ORIGCF502463
Anti-BTRC Mouse Monoclonal Antibody [clone: 1F10]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: BTRC
Clonality: Monoclonal
Clone: 1F10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR