Anti-ZFAND2B Mouse Monoclonal Antibody [clone: 3A5]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF502441EA 5750 DKK
ORIGCF502441
Anti-ZFAND2B Mouse Monoclonal Antibody [clone: 3A5]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: ZFAND2B
Clonality: Monoclonal
Clone: 3A5
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR