Anti-CD4 Mouse Monoclonal Antibody [clone: 4A11]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF500479EA 5750 DKK
ORIGCF500479
Anti-CD4 Mouse Monoclonal Antibody [clone: 4A11]
Antistoffer
Recommended Dilutions: Western Blot: 1:200-1:1000; Immunofluorescence: 1:100

Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: 4A11
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR