Anti-BRAF Mouse Monoclonal Antibody [clone: 3E8]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF500441EA 5590 DKK
ORIGCF500441
Anti-BRAF Mouse Monoclonal Antibody [clone: 3E8]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100

Type: Primary
Antigen: BRAF
Clonality: Monoclonal
Clone: 3E8
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR