Anti-EFNA2 Mouse Monoclonal Antibody [clone: 1C9]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF501425EA 5590 DKK
ORIGCF501425
Anti-EFNA2 Mouse Monoclonal Antibody [clone: 1C9]
Antistoffer
Recommended Dilutions: Western Blot: 1:500-2000; Flow Cytometry: 1:100

Type: Primary
Antigen: EFNA2
Clonality: Monoclonal
Clone: 1C9
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR