Anti-MIOX Mouse Monoclonal Antibody [clone: 5F8]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF501424EA 5750 DKK
ORIGCF501424
Anti-MIOX Mouse Monoclonal Antibody [clone: 5F8]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: MIOX
Clonality: Monoclonal
Clone: 5F8
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR