Anti-TTLL12 Mouse Monoclonal Antibody [clone: 5H3]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF500780EA 5750 DKK
ORIGCF500780
Anti-TTLL12 Mouse Monoclonal Antibody [clone: 5H3]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100

Type: Primary
Antigen: TTLL12
Clonality: Monoclonal
Clone: 5H3
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR