Anti-TYRO3 Mouse Monoclonal Antibody [clone: 9B12]
Leverandør: OriGene
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ORIGCF500421EA 5590 DKK
ORIGCF500421
Anti-TYRO3 Mouse Monoclonal Antibody [clone: 9B12]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100; Immunoprecipitation: 2?g/500?l
Type: Primary
Antigen: TYRO3
Clonality: Monoclonal
Clone: 9B12
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human