Anti-STK3 Mouse Monoclonal Antibody [clone: 4G10]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF500408EA 5590 DKK
ORIGCF500408
Anti-STK3 Mouse Monoclonal Antibody [clone: 4G10]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50

Type: Primary
Antigen: STK3
Clonality: Monoclonal
Clone: 4G10
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Dog, Human, Monkey
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR