Anti-CHEK2 Mouse Monoclonal Antibody [clone: 5C4]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF500398EA 5590 DKK
ORIGCF500398
Anti-CHEK2 Mouse Monoclonal Antibody [clone: 5C4]
Antistoffer
Recommended Dilutions: Western Blot: 1:500-1000; Immunochemistry: 1:150; Immunofluorescence: 1:50-100; Flow Cytometry: 1:100; Immunoprecipitation: 2?g/500?l

Type: Primary
Antigen: CHEK2
Clonality: Monoclonal
Clone: 5C4
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR