Anti-CD2 Mouse Monoclonal Antibody [clone: 9D1]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF500394EA 5590 DKK
ORIGCF500394
Anti-CD2 Mouse Monoclonal Antibody [clone: 9D1]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: CD2
Clonality: Monoclonal
Clone: 9D1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR