Anti-PLK1 Mouse Monoclonal Antibody [clone: 1D4]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF500389EA 5590 DKK
ORIGCF500389
Anti-PLK1 Mouse Monoclonal Antibody [clone: 1D4]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunoprecipitation: 2?g/500?l

Type: Primary
Antigen: PLK1
Clonality: Monoclonal
Clone: 1D4
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR