Anti-COX6A1 Mouse Monoclonal Antibody [clone: 6H8]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF501398EA 5590 DKK
ORIGCF501398
Anti-COX6A1 Mouse Monoclonal Antibody [clone: 6H8]
Antistoffer
Recommended Dilutions: Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: COX6A1
Clonality: Monoclonal
Clone: 6H8
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR