Anti-DGKB Mouse Monoclonal Antibody [clone: 1F9]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF501381EA 5590 DKK
ORIGCF501381
Anti-DGKB Mouse Monoclonal Antibody [clone: 1F9]
Antistoffer
Recommended Dilutions: Western Blot: 1:500-2000

Type: Primary
Antigen: DGKB
Clonality: Monoclonal
Clone: 1F9
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR