Anti-IDO2 Mouse Monoclonal Antibody [clone: 1A4]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF501378EA 5750 DKK
ORIGCF501378
Anti-IDO2 Mouse Monoclonal Antibody [clone: 1A4]
Antistoffer
Recommended Dilutions: Western Blot: 1:500; Flow Cytometry: 1:100

Type: Primary
Antigen: IDO2
Clonality: Monoclonal
Clone: 1A4
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR