Anti-EIF2S1 Mouse Monoclonal Antibody [clone: 5A5]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF501374EA 5590 DKK
ORIGCF501374
Anti-EIF2S1 Mouse Monoclonal Antibody [clone: 5A5]
Antistoffer
Recommended Dilutions: Western Blot: 1:1000-2000; Immunochemistry: 1:50

Type: Primary
Antigen: EIF2S1
Clonality: Monoclonal
Clone: 5A5
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR