Anti-AKR1A1 Mouse Monoclonal Antibody [clone: 6E3]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF500745EA 5590 DKK
ORIGCF500745
Anti-AKR1A1 Mouse Monoclonal Antibody [clone: 6E3]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100 Flow Cytometry: 1:100

Type: Primary
Antigen: AKR1A1
Clonality: Monoclonal
Clone: 6E3
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR