Anti-RNF144B Mouse Monoclonal Antibody [clone: 1E1]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF500712EA 5590 DKK
ORIGCF500712
Anti-RNF144B Mouse Monoclonal Antibody [clone: 1E1]
Antistoffer
Recommended Dilutions: Flow Cytometry: 1:100

Type: Primary
Antigen: RNF144B
Clonality: Monoclonal
Clone: 1E1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR