Anti-DNAJB1 Mouse Monoclonal Antibody [clone: 1H8]
Leverandør: OriGene
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ORIGCF502348EA 5590 DKK
ORIGCF502348
Anti-DNAJB1 Mouse Monoclonal Antibody [clone: 1H8]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100
Type: Primary
Antigen: DNAJB1
Clonality: Monoclonal
Clone: 1H8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human