Anti-SERPINA1 Mouse Monoclonal Antibody [clone: 15H10]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF500375EA 5590 DKK
ORIGCF500375
Anti-SERPINA1 Mouse Monoclonal Antibody [clone: 15H10]
Antistoffer
Recommended Dilutions: Western Blot: 1:500-1000; Immunochemistry: 1:150; Immunoprecipitation: 2?g/500?l

Type: Primary
Antigen: SERPINA1
Clonality: Monoclonal
Clone: 15H10
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR