Anti-GFAP Mouse Monoclonal Antibody [clone: 1B3]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF500342EA 5750 DKK
ORIGCF500342
Anti-GFAP Mouse Monoclonal Antibody [clone: 1B3]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: GFAP
Clonality: Monoclonal
Clone: 1B3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR