Anti-TBXAS1 Mouse Monoclonal Antibody [clone: 1H9]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF501357EA 5590 DKK
ORIGCF501357
Anti-TBXAS1 Mouse Monoclonal Antibody [clone: 1H9]
Antistoffer
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:50; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: TBXAS1
Clonality: Monoclonal
Clone: 1H9
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR