Anti-TTC32 Mouse Monoclonal Antibody [clone: 5B4]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF501339EA 5750 DKK
ORIGCF501339
Anti-TTC32 Mouse Monoclonal Antibody [clone: 5B4]
Antistoffer
Recommended Dilutions: Western Blot: 1:1000; Flow Cytometry: 1:100

Type: Primary
Antigen: TTC32
Clonality: Monoclonal
Clone: 5B4
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR