Anti-PSMA7 Mouse Monoclonal Antibody [clone: 2A7]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF500692EA 5590 DKK
ORIGCF500692
Anti-PSMA7 Mouse Monoclonal Antibody [clone: 2A7]
Antistoffer
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:50

Type: Primary
Antigen: PSMA7
Clonality: Monoclonal
Clone: 2A7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR