Anti-PSMA7 Mouse Monoclonal Antibody [clone: 3F11]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF500691EA 5590 DKK
ORIGCF500691
Anti-PSMA7 Mouse Monoclonal Antibody [clone: 3F11]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50 Flow Cytometry: 1:100

Type: Primary
Antigen: PSMA7
Clonality: Monoclonal
Clone: 3F11
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR