Anti-SORD Mouse Monoclonal Antibody [clone: 2A10]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF500683EA 5590 DKK
ORIGCF500683
Anti-SORD Mouse Monoclonal Antibody [clone: 2A10]
Antistoffer
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:50

Type: Primary
Antigen: SORD
Clonality: Monoclonal
Clone: 2A10
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR