Digital microplate shakers, compact
Leverandør: Thermo Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BARN88880024EA 8130 DKK
BARN88880024
Digital microplate shakers, compact
This microplate shaker is ideal for immunoassay, screening, and magnetic bead applications. It is capable of shaking up to four microplates in one run. The compact design makes it ideal for placing in incubators, chambers, and refrigerators.
  • Digital display for easy viewing
  • Automatic overload shutdown
  • PID control ensures consistent shaking results and precise temperature control
Speed accuracy (at 500 min⁻¹): ±15 min⁻¹
Bestillingsoplysninger: Units are supplied without platforms. Universal platforms and other accessories must be ordered separately.