Turbidity meter, TurbiCheck
Leverandør: Lovibond Tintometer
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
LOVI194200EA 15940 DKK
LOVI194200
Turbidity meter, TurbiCheck
The TurbiCheck allows for turbidity measurement in the field or laboratory.
  • Automatic ranging feature
Designed for regulatory monitoring, process control and field use.
Leveringsinformation: Supplied with three turbidity standards, four batteries, two sample vials and a carrying case.