Black sealing foil for microplates, PeelASeal Foil™
Leverandør: IST Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ISTSIST-109-115TREA 282 DKK
ISTSIST-109-115TR ISTSIST-109-078LR ISTSIST-109-078LS ISTSIST-109-115SR ISTSIST-109-078TR ISTSIST-109-115LR ISTSIST-109-078TS ISTSIST-109-078SR ISTSIST-109-078SS
Black sealing foil for microplates, PeelASeal Foil™
Peelable low fluorescence coated foil for fluorescent and light sensitive assays. Typically for use with black microplates.
  • Non pierceable
  • Seal integrity range: −20 to 80 °C
  • Available sterile or non sterile