Protective hand creams, Ligana® B Oleo-Tec
Leverandør: Faweco Hautschutz
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Ligana® Oleo-Tec
FAWE12984001EA 0 DKK
FAWE12984001
Protective hand creams, Ligana® B Oleo-Tec
Hudpleje Cremer Hand Creams
O/W cream protects from oil-soluble substances such as oils, organic solvents, non-water soluble cooling lubricants, cutting oils, and other oily substances.
  • Reduces skin irritations
  • Contains glycerine and beeswax
  • Silicone-free, perfumed
Advarsel: *Produktet/produkterne på denne side er uden for anvendelsesområdet for EU -direktiv 2019/904