IPA spray, sterile, IPASEPT® 70 Premium
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
94018.1006EA 0 DKK
VWRC94018.1006
IPA spray, sterile, IPASEPT® 70 Premium
70% Iso-propyl alcohol compliant with Ph. Eur. and 30% water for injection according to USP, filtered at 0,2 μm
  • Ready to use, eliminates the need for on-site preparation of alcohol solution
  • 0,2 μm filtered and irradiated with SAL 10⁻⁶, for use in Grade A and B cleanrooms
  • Airless system, preventing airborne contamination being sucked in
  • Collapsible inner pouch, reducing waste and disposal costs
  • Double bagged for easy transfer into a cleanroom
  • Traceability and assurance of quality
IPASEPT 70 Premium is ideal for the routine cleaning of all surfaces in pharmaceutical facilities, such as cleanroom work surfaces, process equipment, isolators and biosafety cabinets, and more. It is also recommended for decontamination of materials before entering the cleanroom, and for spraying on gloved hands.
Emballering: Double bagged and labelled with batch number and shelf life.