Alti, baro and thermometer
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
620-1953EA 1140 DKK
VWRI620-1953
Alti, baro and thermometer
This precision alti, baro and thermometer is ideal for professional use in sports and technology.
  • Lucent-screening grid front (IP 65)
  • Maximum/minimum value memory
  • Tendency meter for falling and/or rising air pressure
  • Sea level adjustment and summing function
  • Over 6000 operating hours
Piezo-resistive absolute pressure and temperature sensor.
Leveringsinformation: Includes 9 volt battery type IEC 6F22 and low battery warning.