PTFE balls, Chemware®
Leverandør: Saint-Gobain
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Chemware®
VERND1069326EA 698 DKK
VERND1069326 VERND1069391
PTFE balls, Chemware®
Kugler Plastikkugler
Chemically resistant balls for packaging of columns and mixing beads. Non-contaminating and minimize bumping during boiling.
  • Inert
  • Temperature restistance: −200 to 288 °C